siRNA Details
| Gene Symbol: Mk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134399.2 | NP_599226.1 | 1 | 2089 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mk1| Gene Symbol: Mk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134399.2 | NP_599226.1 | 1 | 2089 | ||