siRNA Details
| Gene Symbol: LOC257650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147142.1 | NP_671483.1 | 2, 3 | 121 | ||
| GenBank mRNA | AF260582.1 | - | 2, 3 | 121 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC257650| Gene Symbol: LOC257650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147142.1 | NP_671483.1 | 2, 3 | 121 | ||
| GenBank mRNA | AF260582.1 | - | 2, 3 | 121 | ||