siRNA Details
| Gene Symbol: St6gal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147205.2 | NP_671738.2 | 2 | 182 | ||
| GenBank mRNA | M73987.1 | - | 2 | 219 | ||
| M83143.1 | - | 2 | 182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St6gal1| Gene Symbol: St6gal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147205.2 | NP_671738.2 | 2 | 182 | ||
| GenBank mRNA | M73987.1 | - | 2 | 219 | ||
| M83143.1 | - | 2 | 182 | |||