siRNA Details
| Gene Symbol: Elavl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172324.1 | NP_758827.1 | 2 | 157 | ||
| XM_006242616.2 | XP_006242678.1 | 2 | 501 | |||
| XM_008765920.1 | XP_008764142.1 | 2 | 344 | |||
| GenBank mRNA | AB097198.1 | - | 2 | 157 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010487, U29148, BC052097, AK090281.

Elavl3
Genomic Map