siRNA Details
| Gene Symbol: Chmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172331.1 | NP_758834.1 | 2 | 95 | ||
| XM_006236631.2 | XP_006236693.1 | 2 | 204 | |||
| GenBank mRNA | AY150169.1 | - | 2 | 95 | ||
| FQ215758.1 | - | 2 | 109 | |||
| FQ219218.1 | - | 2 | 114 | |||

Chmp3
Genomic Map