siRNA Details
| Gene Symbol: Hmgcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173094.2 | NP_775117.2 | 1, 2 | 133 | ||
| XM_006233002.2 | XP_006233064.1 | 1, 2 | 162 | |||
| GenBank mRNA | BC078695.1 | - | 1, 2 | 133 | 7112644 | |
| BC083543.1 | - | 1, 2 | 133 | 7132009 | ||
| M33648.1 | - | 1, 2 | 151 | |||

Hmgcs2
Genomic Map