siRNA Details
| Gene Symbol: Parm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173114.1 | NP_775137.1 | 2 | 291 | ||
| GenBank mRNA | AJ010750.1 | - | 2 | 313 | ||
| BC060539.1 | - | 2 | 317 | 6890269 | ||
| FQ219991.1 | - | 2 | 314 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Parm1| Gene Symbol: Parm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173114.1 | NP_775137.1 | 2 | 291 | ||
| GenBank mRNA | AJ010750.1 | - | 2 | 313 | ||
| BC060539.1 | - | 2 | 317 | 6890269 | ||
| FQ219991.1 | - | 2 | 314 | |||