siRNA Details
| Gene Symbol: Vimp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173120.2 | NP_775143.2 | 2 | 131 | ||
| GenBank mRNA | AF367467.1 | - | 2 | 141 | ||
| BC059111.1 | - | 2 | 135 | 6920258 | ||
| FQ210095.1 | - | 2 | 123 | |||
| FQ218853.1 | - | 2 | 141 | |||
| FQ219944.1 | - | 2 | 123 | |||
| FQ220082.1 | - | 2 | 126 | |||
| FQ225575.1 | - | 2 | 141 | |||
| FQ229642.1 | - | 2 | 123 | |||

Vimp
Genomic Map