siRNA Details
| Gene Symbol: Cyp4f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173123.1 | NP_775146.1 | 2 | 259 | ||
| GenBank mRNA | U39206.1 | - | 2 | 259 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4f4| Gene Symbol: Cyp4f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173123.1 | NP_775146.1 | 2 | 259 | ||
| GenBank mRNA | U39206.1 | - | 2 | 259 | ||