siRNA Details
Gene Symbol: Testin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173132.2 | NP_775155.1 | 2 | 164 | ||
XM_008771428.1 | XP_008769650.1 | 3 | 524 | |||
GenBank mRNA | U16858.1 | - | 2 | 168 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Testin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173132.2 | NP_775155.1 | 2 | 164 | ||
XM_008771428.1 | XP_008769650.1 | 3 | 524 | |||
GenBank mRNA | U16858.1 | - | 2 | 168 |