siRNA Details
| Gene Symbol: Pex5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173152.1 | NP_775175.1 | 134 | |||
| GenBank mRNA | AF324454.1 | - | 134 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001163516, NM_021483, HQ646194, HQ646192, HQ646188, and others.

Pex5l
Genomic Map