siRNA Details
| Gene Symbol: Cryge | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173289.1 | NP_775411.1 | 3 | 307 | ||
| XM_008767178.1 | XP_008765400.1 | 4 | 484 | |||
| GenBank mRNA | J00716.1 | - | 265 | |||
| Gene Symbol: Crygf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109557.1 | NP_001103027.1 | 3 | 360 | ||

Cryge
Genomic Map