siRNA Details
Gene Symbol: Vom1r97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173297.2 | NP_775419.2 | 1 | 222 | ||
GenBank mRNA | AY510281.1 | - | 1 | 222 | ||
U36897.1 | - | 1 | 128 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vom1r97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173297.2 | NP_775419.2 | 1 | 222 | ||
GenBank mRNA | AY510281.1 | - | 1 | 222 | ||
U36897.1 | - | 1 | 128 |