siRNA Details
| Gene Symbol: Hsd17b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173305.1 | NP_775427.1 | 2 | 335 | ||
| XM_006240739.2 | XP_006240801.2 | 2 | 131 | |||
| GenBank mRNA | BC088104.1 | - | 2 | 285 | 7326228 | |
| BC091114.1 | - | 2 | 247 | 7321553 | ||
| U89280.1 | - | 2 | 335 | |||
| Gene Symbol: LOC100362350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769893.1 | XP_008768115.1 | 1 | 117 | ||

Hsd17b6
Genomic Map