siRNA Details
Gene Symbol: Duox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153739.1 | NP_714961.1 | 3 | 209 | ||
XM_006234829.2 | XP_006234891.1 | 4 | 308 | |||
GenBank mRNA | AF542180.1 | - | 3 | 209 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Duox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153739.1 | NP_714961.1 | 3 | 209 | ||
XM_006234829.2 | XP_006234891.1 | 4 | 308 | |||
GenBank mRNA | AF542180.1 | - | 3 | 209 |