siRNA Details
| Gene Symbol: Cthrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172333.2 | NP_758836.1 | 1, 2 | 262 | ||
| GenBank mRNA | AY136824.1 | - | 1, 2 | 264 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cthrc1| Gene Symbol: Cthrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172333.2 | NP_758836.1 | 1, 2 | 262 | ||
| GenBank mRNA | AY136824.1 | - | 1, 2 | 264 | ||