siRNA Details
Gene Symbol: Bdkrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270713.1 | NP_001257642.1 | 4 | 557 | ||
NM_173100.2 | NP_775123.2 | 3 | 383 | |||
GenBank mRNA | M59967.1 | - | 4 | 551 | ||
X69681.1 | - | 3 | 383 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
