siRNA Details
| Gene Symbol: Cyp4f5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173124.1 | NP_775147.1 | 179 | |||
| XM_006241051.2 | XP_006241113.1 | 2 | 248 | |||
| XM_008765157.1 | XP_008763379.1 | 2 | 250 | |||
| XM_008765158.1 | XP_008763380.1 | 2 | 250 | |||
| GenBank mRNA | BC129085.1 | - | 2 | 140 | 7375658 | |
| U39207.1 | - | 179 | ||||
| Gene Symbol: Cyp4f37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271352.1 | NP_001258281.1 | 1 | 106 | ||

Cyp4f5
Genomic Map