siRNA Details
Gene Symbol: Tfpi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173141.1 | NP_775164.1 | 2 | 213 | ||
GenBank mRNA | AJ428954.1 | - | 2 | 213 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tfpi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173141.1 | NP_775164.1 | 2 | 213 | ||
GenBank mRNA | AJ428954.1 | - | 2 | 213 |