siRNA Details
| Gene Symbol: Dlgap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173145.1 | NP_775168.1 | 2 | 539 | ||
| GenBank mRNA | U67140.1 | - | 2 | 539 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlgap4| Gene Symbol: Dlgap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173145.1 | NP_775168.1 | 2 | 539 | ||
| GenBank mRNA | U67140.1 | - | 2 | 539 | ||