siRNA Details
| Gene Symbol: Pex5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173152.1 | NP_775175.1 | 153 | |||
| GenBank mRNA | AF324454.1 | - | 153 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pex5l| Gene Symbol: Pex5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173152.1 | NP_775175.1 | 153 | |||
| GenBank mRNA | AF324454.1 | - | 153 | |||