siRNA Details
Gene Symbol: Slc15a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139341.1 | NP_647557.1 | 5 | 1258 | ||
GenBank mRNA | AB026665.1 | - | 5 | 1258 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc15a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139341.1 | NP_647557.1 | 5 | 1258 | ||
GenBank mRNA | AB026665.1 | - | 5 | 1258 |