siRNA Details
Gene Symbol: Gpr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153727.1 | NP_714949.1 | 1 | 422 | ||
XM_006239006.2 | XP_006239068.1 | 2 | 561 | |||
GenBank mRNA | AJ427482.1 | - | 1 | 422 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
