siRNA Details
| Gene Symbol: Vimp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF367467.1 | - | 3, 4 | 349 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vimp| Gene Symbol: Vimp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF367467.1 | - | 3, 4 | 349 | ||