siRNA Details
| Gene Symbol: Akr1b8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173136.1 | NP_775159.1 | 3 | 305 | ||
| GenBank mRNA | AJ277957.1 | - | 3 | 305 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akr1b8| Gene Symbol: Akr1b8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173136.1 | NP_775159.1 | 3 | 305 | ||
| GenBank mRNA | AJ277957.1 | - | 3 | 305 | ||