siRNA Details
| Gene Symbol: Cryge | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173289.1 | NP_775411.1 | 3 | 556 | ||
| XM_008767178.1 | XP_008765400.1 | 4 | 733 | |||
| GenBank mRNA | J00716.1 | - | 514 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cryge| Gene Symbol: Cryge | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173289.1 | NP_775411.1 | 3 | 556 | ||
| XM_008767178.1 | XP_008765400.1 | 4 | 733 | |||
| GenBank mRNA | J00716.1 | - | 514 | |||