siRNA Details
Gene Symbol: Olr1271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173300.1 | NP_775422.1 | 2 | 1136 | ||
GenBank mRNA | X80671.1 | - | 2 | 1136 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173300.1 | NP_775422.1 | 2 | 1136 | ||
GenBank mRNA | X80671.1 | - | 2 | 1136 |