siRNA Details
| Gene Symbol: Cyp2d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173304.2 | NP_775426.1 | 4 | 668 | ||
| GenBank mRNA | BC088097.1 | - | 4 | 670 | 7369436 | |
| J02869.1 | - | 4 | 668 | |||
| M22329.1 | - | 4 | 602 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2d5| Gene Symbol: Cyp2d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173304.2 | NP_775426.1 | 4 | 668 | ||
| GenBank mRNA | BC088097.1 | - | 4 | 670 | 7369436 | |
| J02869.1 | - | 4 | 668 | |||
| M22329.1 | - | 4 | 602 | |||