siRNA Details
| Gene Symbol: Ceacam10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173339.1 | NP_775461.1 | 2 | 297 | ||
| GenBank mRNA | U23056.1 | - | 2 | 297 | ||
| Gene Symbol: LOC103689942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759077.1 | XP_008757299.1 | 2 | 331 | ||

Ceacam10
Genomic Map