siRNA Details
| Gene Symbol: Ncstn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174864.3 | NP_777353.1 | 2 | 203 | ||
| XM_006250283.1 | XP_006250345.1 | 2 | 182 | |||
| XM_008769733.1 | XP_008767955.1 | 2 | 188 | |||
| XM_008769734.1 | XP_008767956.1 | 1 | 30 | |||
| XM_008769735.1 | XP_008767957.1 | 1 | 30 | |||
| GenBank mRNA | AF510722.1 | - | 2 | 111 | ||
| AY101378.1 | - | 2 | 111 | |||
| BC074006.1 | - | 2 | 203 | |||
| BC167747.1 | - | 2 | 182 | 8365089 | ||

Ncstn
Genomic Map