siRNA Details
Gene Symbol: Cacna2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175595.2 | NP_783185.1 | 2 | 164 | ||
GenBank mRNA | AF486278.1 | - | 2 | 164 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cacna2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175595.2 | NP_783185.1 | 2 | 164 | ||
GenBank mRNA | AF486278.1 | - | 2 | 164 |