siRNA Details
Gene Symbol: Ldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175762.2 | NP_786938.1 | 1, 2 | 228 | ||
GenBank mRNA | X13722.1 | - | 1, 2 | 228 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175762.2 | NP_786938.1 | 1, 2 | 228 | ||
GenBank mRNA | X13722.1 | - | 1, 2 | 228 |