siRNA Details
Gene Symbol: Cpg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178104.2 | NP_835205.1 | 2 | 981 | ||
GenBank mRNA | X73292.1 | - | 2 | 981 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cpg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178104.2 | NP_835205.1 | 2 | 981 | ||
GenBank mRNA | X73292.1 | - | 2 | 981 |