siRNA Details
| Gene Symbol: Mapk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173331.2 | NP_775453.1 | 2, 3 | 216 | ||
| XM_006241777.2 | XP_006241839.1 | 3, 4 | 632 | |||
| XM_006241779.2 | XP_006241841.1 | 3, 4 | 632 | |||
| XM_008765547.1 | XP_008763769.1 | 3, 4 | 484 | |||
| GenBank mRNA | AF078798.2 | - | 2, 3 | 332 | ||

Mapk15
Genomic Map