siRNA Details
| Gene Symbol: Cacna2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175592.2 | NP_783182.1 | 2, 3 | 296 | ||
| XM_006243734.2 | XP_006243796.1 | 2, 3 | 570 | |||
| XM_006243735.2 | XP_006243797.1 | 2, 3 | 570 | |||
| XM_006243736.2 | XP_006243798.1 | 2, 3 | 570 | |||
| XM_006243737.2 | XP_006243799.1 | 2, 3 | 570 | |||
| GenBank mRNA | AF486277.1 | - | 2, 3 | 296 | ||

Cacna2d2
Genomic Map