siRNA Details
| Gene Symbol: Psip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175765.2 | NP_786941.1 | 2 | 202 | ||
| XM_006238353.2 | XP_006238415.1 | 3 | 426 | |||
| XM_006238354.2 | XP_006238416.1 | 3 | 425 | |||
| XM_008763779.1 | XP_008762001.1 | 3 | 425 | |||
| GenBank mRNA | AB285525.1 | - | 105 | |||
| AF339084.1 | - | 2 | 202 | |||
| AF372090.1 | - | 2 | 202 | |||
| BC093606.1 | - | 3 | 360 | 7317952 | ||

Psip1
Genomic Map