siRNA Details
| Gene Symbol: Mtus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB190507.1 | - | 2 | 517 | ||
| AY208915.1 | - | 2 | 208 | |||
| BC166591.1 | - | 2 | 489 | 7938925 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtus1| Gene Symbol: Mtus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB190507.1 | - | 2 | 517 | ||
| AY208915.1 | - | 2 | 208 | |||
| BC166591.1 | - | 2 | 489 | 7938925 | ||