siRNA Details
| Gene Symbol: Itpkc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178094.3 | NP_835195.1 | 1 | 440 | ||
| GenBank mRNA | AJ440782.1 | - | 1 | 440 | ||
| AY160770.1 | - | 1 | 686 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itpkc| Gene Symbol: Itpkc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178094.3 | NP_835195.1 | 1 | 440 | ||
| GenBank mRNA | AJ440782.1 | - | 1 | 440 | ||
| AY160770.1 | - | 1 | 686 | |||