siRNA Details
| Gene Symbol: Cpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178104.2 | NP_835205.1 | 2 | 984 | ||
| GenBank mRNA | X73292.1 | - | 2 | 984 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpg1| Gene Symbol: Cpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178104.2 | NP_835205.1 | 2 | 984 | ||
| GenBank mRNA | X73292.1 | - | 2 | 984 | ||