siRNA Details
| Gene Symbol: Igf2bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175594.2 | NP_783184.1 | 14 | 1844 | ||
| XM_006247193.2 | XP_006247255.1 | 14 | 2162 | |||
| GenBank mRNA | AF541940.1 | - | 14 | 1844 | ||
| BC161831.1 | - | 14 | 1876 | 9023972 | ||

Igf2bp1
Genomic Map