siRNA Details
| Gene Symbol: Hsp90aa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175761.2 | NP_786937.1 | 7 | 1300 | ||
| GenBank mRNA | AJ428213.1 | - | 7 | 1300 | ||
| BC072489.1 | - | 7 | 1282 | 7105514 | ||
| BC085120.1 | - | 7 | 1282 | 7114546 | ||
| Gene Symbol: LOC103692716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764969.1 | XP_008763191.1 | 1352 | |||

Hsp90aa1
Genomic Map