siRNA Details
| Gene Symbol: Kcnmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176861.1 | NP_789831.1 | 3 | 408 | ||
| GenBank mRNA | AJ517198.1 | - | 2 | 292 | ||
| AY191836.1 | - | 3 | 408 | |||
| BC081695.1 | - | 708 | 7190722 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnmb2| Gene Symbol: Kcnmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176861.1 | NP_789831.1 | 3 | 408 | ||
| GenBank mRNA | AJ517198.1 | - | 2 | 292 | ||
| AY191836.1 | - | 3 | 408 | |||
| BC081695.1 | - | 708 | 7190722 | |||