siRNA Details
| Gene Symbol: Serpinf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177927.2 | NP_808788.1 | 7 | 864 | ||
| GenBank mRNA | AY216659.1 | - | 7 | 864 | ||
| BC078686.1 | - | 7 | 852 | 7110230 | ||
| FQ218413.1 | - | 7 | 865 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinf1| Gene Symbol: Serpinf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177927.2 | NP_808788.1 | 7 | 864 | ||
| GenBank mRNA | AY216659.1 | - | 7 | 864 | ||
| BC078686.1 | - | 7 | 852 | 7110230 | ||
| FQ218413.1 | - | 7 | 865 | |||