siRNA Details
| Gene Symbol: Sel1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177933.2 | NP_808794.2 | 6 | 826 | ||
| GenBank mRNA | AF304853.1 | - | 6 | 840 | ||
| AF304855.1 | - | 2 | 225 | |||
| CB814385.1 | - | 3 | 261 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sel1l| Gene Symbol: Sel1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177933.2 | NP_808794.2 | 6 | 826 | ||
| GenBank mRNA | AF304853.1 | - | 6 | 840 | ||
| AF304855.1 | - | 2 | 225 | |||
| CB814385.1 | - | 3 | 261 | |||