siRNA Details
| Gene Symbol: Abca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178095.2 | NP_835196.1 | 4, 5 | 543 | ||
| XM_008763708.1 | XP_008761930.1 | 7, 8 | 1126 | |||
| XM_008763709.1 | XP_008761931.1 | 7, 8 | 1126 | |||
| XM_008763710.1 | XP_008761932.1 | 7, 8 | 1125 | |||
| GenBank mRNA | AY208182.1 | - | 4, 5 | 543 | ||

Abca1
Genomic Map