siRNA Details
| Gene Symbol: Gpm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178105.2 | NP_835206.1 | 7 | 779 | ||
| XM_006253093.2 | XP_006253155.1 | 8 | 1615 | |||
| GenBank mRNA | AB089242.1 | - | 7 | 758 | ||
| BC088862.1 | - | 7 | 779 | 7313006 | ||

Gpm6a
Genomic Map