siRNA Details
| Gene Symbol: Adrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012492.2 | NP_036624.2 | 1 | 768 | ||
| GenBank mRNA | AY057895.1 | - | 1016 | |||
| AY057896.1 | - | 1 | 1017 | |||
| BC086538.1 | - | 1 | 768 | 7114502 | ||
| J03024.1 | - | 1 | 670 | |||
More Information
Set Membership: |
|

Adrb2
Genomic Map