siRNA Details
| Gene Symbol: Apc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012499.1 | NP_036631.1 | 1, 2 | 181 | ||
| XM_006254508.2 | XP_006254570.1 | 2, 3 | 228 | |||
| XM_006254509.2 | XP_006254571.1 | 2, 3 | 197 | |||
| XM_006254511.2 | XP_006254573.1 | 2, 3 | 228 | |||
| XM_006254521.2 | XP_006254583.1 | 2, 3 | 228 | |||
| GenBank mRNA | D38629.1 | - | 1, 2 | 181 | ||

Apc
Genomic Map