siRNA Details
Gene Symbol: Ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012502.1 | NP_036634.1 | 1 | 1586 | ||
GenBank mRNA | M20133.1 | - | 1 | 1586 | ||
M23264.1 | - | 1 | 625 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012502.1 | NP_036634.1 | 1 | 1586 | ||
GenBank mRNA | M20133.1 | - | 1 | 1586 | ||
M23264.1 | - | 1 | 625 |