siRNA Details
| Gene Symbol: Atp1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012506.1 | NP_036638.1 | 2, 3 | 232 | ||
| XM_008758888.1 | XP_008757110.1 | 2, 3 | 146 | |||
| GenBank mRNA | FQ212331.1 | - | 1, 2 | 69 | ||
| M14513.1 | - | 2, 3 | 232 | |||
| M28648.1 | - | 1, 2 | 89 | |||
| X05883.1 | - | 2, 3 | 229 | |||

Atp1a3
Genomic Map